missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Stearic acid, 97%, Thermo Scientific Chemicals
$83.59 - $422.41
Chemical Identifiers
CAS | 57-11-4 |
---|---|
Molecular Formula | C18H36O2 |
Molecular Weight (g/mol) | 284.48 |
MDL Number | MFCD00002752 |
InChI Key | QIQXTHQIDYTFRH-UHFFFAOYSA-N |
Synonym | stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid |
PubChem CID | 5281 |
ChEBI | CHEBI:28842 |
IUPAC Name | octadecanoic acid |
SMILES | CCCCCCCCCCCCCCCCCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC174490010
|
Thermo Scientific Chemicals
174490010 |
1 kg | Plastic Bottle |
Each for $83.59
|
|
|||||
AC174490100
|
Thermo Scientific Chemicals
174490100 |
10 kg | Fibre drum |
Each for $422.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57-11-4 | |
284.48 | |
QIQXTHQIDYTFRH-UHFFFAOYSA-N | |
5281 | |
octadecanoic acid |
C18H36O2 | |
MFCD00002752 | |
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid | |
CHEBI:28842 | |
CCCCCCCCCCCCCCCCCC(O)=O |
Specifications
57-11-4 | |
White | |
361.0°C | |
Authentic | |
Plastic Bottle | |
CH3(CH2)16CO2H | |
1 kg | |
0.84 | |
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid | |
Solubility in water: slightly soluble. Other solubilities: 1g/21mL alcohol- 1g/6mL ccl4,1g/5mL benzene- 1g/3.4mL cs2,1g/2mL chloroform - 1g/26mL acetone,soluble in amyl acetate and toluene | |
CCCCCCCCCCCCCCCCCC(O)=O | |
284.48 | |
CHEBI:28842 | |
97% | |
Stearic acid |
67.0°C to 69.0°C | |
0.8400g/mL | |
196°C | |
96% min. (GC) | |
C18H36O2 | |
MFCD00002752 | |
02,IV,1206 | |
15,893 | |
>90 °C | |
QIQXTHQIDYTFRH-UHFFFAOYSA-N | |
octadecanoic acid | |
5281 | |
284.47 | |
Granular Powder, Granules, or Flakes |
Safety and Handling
EINECSNumber : 200-313-4
RUO – Research Use Only