Fisherbrand™ Steel Mobile Cabinet
Fisherbrand™ Steel Mobile cabinets on locking swivel casters are ideal for under a table or workstation lab environment.
 View more versions of this product
Catalog No. 06000352
Explore available promotions
Description
Description
  • Cabinet has a finished back and top
  • Provided without locks
  • Counter weighted anti-tip design
  • Optional Work surface sold separately
Compliance
SEFA 8
Specifications
Specifications
Locking swivel casters and drawer interlock system
3
22 in.
55.9 cm
32.125 in.
White
18 in.
Mobile Cabinet
81.6 cm
45.7 cm
Mobile
Steel