Fisherbrand™ Steel Mobile Cabinet
Fisherbrand™ Steel Mobile cabinets are on locking swivel casters which allow them to be moved. Ideal for under a table or workstation lab environment.
 View more versions of this product
Catalog No. 06003265
Explore available promotions
Description
Description
  • Cabinet has a finished back and top
  • Provided without locks
  • Counter weighted anti-tip design
  • Optional Work surface sold separately
Compliance
SEFA 8
Specifications
Specifications
Locking Swivel Casters
1
22 in.
55.9 cm
27.065 in.
White
24 in.
Mobile Cabinet
68.75 cm
61 cm
1
Steel