missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Steel Wash Tanks

Constructed of lead-free coated steel

$627.85 - $675.57

Specifications

Product Type Wash Tank
Certifications/Compliance FM, TUV
Color Red
For Use With (Application) For Small Parts Cleaning
Material Steel
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (English) Includes Price Quantity  
Catalog Number Mfr. No. Capacity (English) Includes Price Quantity  
17-985-84B
View Documents Promotion Details
Justrite™
JMC27716
6 gal. Fusible link, self close cover and basket
Each for $675.57
 
17-985-84A
View Documents Promotion Details
Justrite™
JMC27713
3.5 gal. Fusible link, self close cover and basket
Each for $627.85
 
Description

Description

  • Cover opens wide for easy access; integral basket facilitates washing of many small parts quickly
  • After cleaning, place basket at top of tank to drain solvent
  • Fusible link melts at 165° F (74° C) and slams the cover shut in case of fire
  • 3.5 gal. size is also available with replaceable polyethylene liner in the top and tank for use with harsh, corrosive chemicals

Certifications

All models FM approved.

Specifications

Specifications

Wash Tank
Red
Steel
FM, TUV
For Small Parts Cleaning
SDS
missing translation for 'documents'

missing translation for 'documents'