Fisherbrand™ Round-Bottom Polystyrene Test Tubes With Cap
Fisherbrand™'s round bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
$130.13 - $531.43
Specifications
Color | Clear |
---|---|
Material | Polystyrene |
Disposable | Single-use |
Product Line | Fisherbrand |
Graduated | No |
Catalog Number | Mfr. No. | Sterility | Capacity (Metric) | Closure Type | Packaging | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sterility | Capacity (Metric) | Closure Type | Packaging | Price | Quantity | ||
FB149563B
|
N/A | Sterile | 5 mL | Snap Cap | Individually wrapped |
Case of 500 Each for $132.43
|
|
||
FB1496120
|
N/A | Non-sterile | 5 mL | Snap Cap | Bulk |
Case of 1000 Each for $165.04
|
|
||
FB149563D
|
N/A | Sterile | 5 mL | Snap Cap | Bag (8 Pk. of 125 Tubes Ea.) |
Case of 1000 Each for $191.61
|
|
||
FB149563C
|
N/A | Sterile | 5 mL | Snap Cap | Blister Pk. (20 Pk. of 25 Tubes Ea.) |
Case of 500 Each for $130.13
|
|
||
FB149564A
|
N/A | Sterile | 8 mL | Screw Cap | Blister Pk. |
Case of 1000 Each for $330.00
|
|
||
FB149566B
|
N/A | Sterile | 14 mL | Snap Cap | Blister Pk. (8 Pk. of 125 Tubes Ea.) |
Case of 1000 Each for $413.09
|
|
||
FB149566D
|
N/A | Sterile | 14 mL | Snap Cap | Blister Pk. (20 Pk. of 25 Tubes Ea.) |
Case of 500 Each for $175.65
|
|
||
FB149566A
|
N/A | Sterile | 14 mL | Snap Cap | Individually wrapped |
Case of 500 Each for $235.96
|
|
||
FB149565A
|
N/A | Sterile | 16 mL | Screw Cap | Individually Wrapped |
Case of 500 Each for $236.74
|
|
||
FB149565B
|
N/A | Sterile | 16 mL | Screw Cap | Blister Pk. |
Case of 1000 Each for $531.43
|
|
||
FB149565C
|
N/A | Sterile | 19 mL | Screw Cap | Individually Wrapped |
Case of 500 Each for $284.39
|
|
||
Description
- Ideal for use in laboratory protocols requiring round bottom tubes
- Ensures reliable handling of liquid samples
- Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
- Available with and without snap cap - dual-position snap caps offer both vented and fully closed positions.
- 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
- Dimensions and volumes are approximate
Specifications
Clear | |
Single-use | |
No |
Polystyrene | |
Fisherbrand | |
Test Tube |