missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Air-Tite™ Sterile Syringes
Air-Tite™ Sterile Syringes are high quality, yet economical for laboratory and veterinary use only
$26.62 - $57.66
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | Clear |
For Use With (Application) | Laboratory / Veterinary |
Latex Free | Yes |
Disposable | Single-use |
Catalog Number | Mfr. No. | Product Type | Volume (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Volume (Metric) | Price | Quantity | ||||
14-817-179
|
Air-Tite™
MS1 |
Luer Slip | 1 mL |
Pack of 100 Each for $26.62
Case of 10 Pack for $266.20
|
|
||||
14-817-108
|
Air-Tite™
ML1 |
Luer Lock | 1 mL |
Pack of 100 Each for $57.66
Case of 10 Pack for $576.60
|
|
||||
14-817-109
|
Air-Tite™
ML3 |
Luer Lock | 3 mL |
Pack of 100 Each for $29.10
Case of 10 Pack for $291.00
|
|
||||
14-817-180
|
Air-Tite™
MS5 |
Luer Slip | 5 mL |
Pack of 100 Each for $35.21
Case of 10 Pack for $352.10
|
|
||||
14-817-181
|
Air-Tite™
MS10 |
Luer Slip | 10 mL |
Pack of 100 Each for $46.52
|
|
||||
14-817-111
|
Air-Tite™
AIRML20 |
Luer Lock | 20 mL |
Pack of 50 Each for $48.21
|
|
||||
14-817-112
|
Air-Tite™
ML30 |
Luer Lock | 30 mL |
Pack of 50 Each for $51.48
Case of 10 Pack for $514.80
|
|
||||
14-817-178
|
Air-Tite™
ML10 |
Luer Lock | 10 mL |
Pack of 100 Each for $37.26
Case of 10 Pack for $372.60
|
|
||||
14-817-113
|
Air-Tite™
ML60 |
Luer Lock | 60 mL |
Pack of 25 Each for $45.28
Case of 10 Pack for $452.80
|
|
||||
Description
- Smooth plunger
- Bold graduations
- Disposable
- Sterile
- Polypropylene
- Economical
- Single-use
- Latex Free
- ISO Compatible
Specifications
Not Autoclavable | |
Laboratory / Veterinary | |
Single-use | |
Yes |
Clear | |
Yes | |
Individually Wrapped | |
Sterile |
Not for human use - laboratory and veterinary use only