missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Air-Tite™ Sterile Syringes with Needles - Luer Lock

Air-Tite™ Sterile Syringes with Needles are high quality, yet economical for laboratory and veterinary use only

Supplier:  Air-Tite™ ML520112

 View more versions of this product

Catalog No. 14-817-141



Explore available promotions

Description

Description

  • Disposable Syringes with Needle
  • Smooth plunger
  • Bold graduations
  • Sterile
  • Polypropylene
  • Economical
  • Single-use
  • Latex Free
  • ISO
  • Lab/Vet
Specifications

Specifications

Not Autoclavable
Polyisoprene Rubber (Tip), Polypropylene
20 G
Yes
Luer Lock
0.2 mL
Individually Wrapped
Yes
Meets ISO Requirements
Clear
Laboratory / Veterinary
1.5 in.
Single-use
100/Box
5 mL
Sterile
Videos
SDS
Documents

Documents

Promotions

Promotions

Not for human use - laboratory and veterinary use only