Bel-Art™ Sterileware™ Sterile Styrene Sampler Spoons
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sterile Styrene Sampler Spoons are designed to gather small amounts of sample material.
$167.94 - $1063.42
Specifications
Product Type Spoon
Material Polystyrene
Includes Certificate of sterilization
Handle Type Long
Disposable Single-use
View More Specs
Products 8
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Quantity Price Quantity  
14-429D
View Documents
Bel-Art™
H369460000
14.79 mL 200/Cs.
Case of 200 Each for $903.61
03-990-229
View Documents
Bel-Art™
H369400010
1.23 mL 10/Pk.
Case of 10 Each for $198.12
14-429B
View Documents
Bel-Art™
H369420000
2.46 mL 200/Cs.
Case of 200 Each for $1,063.42
03-990-232
View Documents
Bel-Art™
H369460010
14.79 mL 10/Pk.
Case of 10 Each for $167.94
14-429A
View Documents
Bel-Art™
H369400000
1.23 mL 200/Cs.
Case of 200 Each for $1,025.35
03990230
View Documents
Bel-Art™
H369420010
2.46 mL 10/Pk.
Case of 10 Each for $194.00
03-990-231
View Documents
Bel-Art™
H369440010
4.93 mL 10/Pk.
Case of 10 Each for $182.41
14-429C
View Documents
Bel-Art™
H369440000
4.93 mL 200/Cs.
Case of 200 Each for $976.94
Description
Description
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sterile Styrene Sampler Spoons are designed to gather small amounts of sample material.
  • Sterile: individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
  • FDA grade materials; safe for food, drug and cosmetic contact
  • Long handle reaches easily into jars, bottles and othe confined areas, and minimizes the potential for personal contact with sampled material
  • Overall length 17.8cm (7 in.)
Specifications
Specifications
Spoon
Certificate of sterilization
Single-use
White
Designed to gather small amounts of sample material
17.8 cm
Polystyrene
Long
Sterile
FDA
7 in.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'