missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Streptomycin Sulfate, White Powder, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210055680

 View more versions of this product

Catalog No. ICN10055680

Explore available promotions

Description

Description

Specifications

Specifications

Streptomycin Sulfate
Solid
White
C42H84N14O36S3
Streptomycin Sesquisulfate, Streptomycin A
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
1457.376
1457.38
Streptomycin Sulfate
3810-74-0
100 g
MFCD00037023
QTENRWWVYAAPBI-FFCQDDOVSA-N
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
124080941

Chemical Identifiers

3810-74-0
1457.376
QTENRWWVYAAPBI-FFCQDDOVSA-N
124080941
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
C42H84N14O36S3
MFCD00037023
Streptomycin Sesquisulfate, Streptomycin A
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C.

SDS
Documents

Documents

Promotions

Promotions