missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Streptomycin Sulfate, White Powder, MP Biomedicals™

$58.68 - $868.55

Chemical Identifiers

CAS 3810-74-0
Molecular Formula C42H84N14O36S3
Molecular Weight (g/mol) 1457.376
MDL Number MFCD00037023
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
Synonym Streptomycin Sesquisulfate, Streptomycin A
PubChem CID 124080941
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10055605
View Documents
MP Biomedicals Inc
0210055605
5 g
Each for $58.68
 
ICN10055650
View Documents
MP Biomedicals Inc
0210055650
50 g
Each for $85.38
 
ICN10055680
View Documents
MP Biomedicals Inc
0210055680
100 g
Each for $129.96
 
ICN10055691
View Documents
MP Biomedicals Inc
0210055691
1 kg
Each for $868.55
 
Description

Description

Specifications

Chemical Identifiers

3810-74-0
1457.376
QTENRWWVYAAPBI-FFCQDDOVSA-N
124080941
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
C42H84N14O36S3
MFCD00037023
Streptomycin Sesquisulfate, Streptomycin A
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(

Specifications

Streptomycin Sulfate
3810-74-0
5 g
MFCD00037023
QTENRWWVYAAPBI-FFCQDDOVSA-N
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
124080941
Streptomycin Sulfate
Solid
White
C42H84N14O36S3
Streptomycin Sesquisulfate, Streptomycin A
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
1457.376
1457.38
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C.

SDS
missing translation for 'documents'

missing translation for 'documents'