missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals

Strontium nitrate, 99+%, N2O6Sr, CAS Number-10042-76-9, bdg873aqzl, strontium dinitrate, nitric acid, strontium salt 2:1, hsdb 787, nitric acid, strontium salt, nitrate de strontium french, strontium nitrate sr no3 2, strontium nitrate, strontium ii nitrate 1:2, unii-bdg873aqzl, 1kg

Supplier:  Thermo Scientific Chemicals 222562500

 View more versions of this product

Catalog No. AC222562500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Strontium nitrate
p.a.
White
Crystalline Powder and/or Crystals
99% min. (Complexometry)
Sr(NO3)2
15, 8975
Solubility in water: 660g/L (20°C). Other solubilities: slightly soluble in alcohol and acetone
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
211.63
211.63
Analytical
Plastic bottle
570.0°C
10042-76-9
5 to 7 (5% soln. at 20°C)
250 g
N2O6Sr
MFCD00011248
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium;dinitrate
24848
≥99%
0.1% max. (105°C)
2.99

Chemical Identifiers

10042-76-9
211.63
DHEQXMRUPNDRPG-UHFFFAOYSA-N
24848
N2O6Sr
MFCD00011248
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 233-131-9

RTECSNumber : WK9800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only