missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals

Strontium nitrate, 99+%, N2O6Sr, CAS Number-10042-76-9, bdg873aqzl, strontium dinitrate, nitric acid, strontium salt 2:1, hsdb 787, nitric acid, strontium salt, nitrate de strontium french, strontium nitrate sr no3 2, strontium nitrate, strontium ii nitrate 1:2, unii-bdg873aqzl, 1kg

$151.70 - $349.69

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC222562500
SDS View Documents
Thermo Scientific Chemicals
222562500
250 g Plastic bottle
Each for $151.70
 
AC222560010
SDS View Documents
Thermo Scientific Chemicals
222560010
1 kg Plastic bottle
Each for $349.69
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10042-76-9
211.63
DHEQXMRUPNDRPG-UHFFFAOYSA-N
24848
N2O6Sr
MFCD00011248
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

570.0°C
White
Crystalline Powder and/or Crystals
99% min. (Complexometry)
Sr(NO3)2
15, 8975
Solubility in water: 660g/L (20°C). Other solubilities: slightly soluble in alcohol and acetone
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
211.63
211.63
Analytical
Plastic bottle
Strontium nitrate, p.a.
10042-76-9
5 to 7 (5% soln. at 20°C)
250 g
N2O6Sr
MFCD00011248
strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
DHEQXMRUPNDRPG-UHFFFAOYSA-N
strontium;dinitrate
24848
≥99%
0.1% max. (105°C)
2.99
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 233-131-9

RTECSNumber : WK9800000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only