missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 132790010

 View more versions of this product

Catalog No. AC132790010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Styrene
100-42-5,98-29-3
Authentic
Glass Bottle
1.5460 to 1.5480 (20°C, 589nm)
MFCD00008612,MFCD00084450
05, 474
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
C=CC1=CC=CC=C1
104.15
7501
104.15
≥98.5%
Liquid
Stabilized
0.9060g/mL
99%
C8H8
C6H5CH=CH2
1 L
15, 8990
PPBRXRYQALVLMV-UHFFFAOYSA-N
styrene
10 to 20ppm p-tert-butylcatechol
CHEBI:27452
7mbar at 20°C
Extra Pure

Chemical Identifiers

100-42-5
104.15
PPBRXRYQALVLMV-UHFFFAOYSA-N
7501
styrene
C8H8
MFCD00008612,MFCD00084450
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
CHEBI:27452
C=CC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only