Learn More
Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals
$78.29 - $888.19
Chemical Identifiers
CAS | 100-42-5 |
---|---|
Molecular Formula | C8H8 |
Molecular Weight (g/mol) | 104.15 |
MDL Number | MFCD00008612,MFCD00084450 |
InChI Key | PPBRXRYQALVLMV-UHFFFAOYSA-N |
Synonym | ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene |
PubChem CID | 7501 |
ChEBI | CHEBI:27452 |
IUPAC Name | styrene |
SMILES | C=CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC132790100
|
Thermo Scientific Chemicals
132790100 |
10 mL | Glass Bottle |
Each for $78.29
|
|
||||
AC132790010
|
Thermo Scientific Chemicals
132790010 |
1 L | Glass Bottle |
Each for $88.16
|
|
||||
AC132790025
|
Thermo Scientific Chemicals
132790025 |
2.5 L | Glass Bottle |
Each for $148.14
Case of 4 Each for $0.00
|
|
||||
AC132790250
|
Thermo Scientific Chemicals
132790250 |
25 L | Metal drum |
Each for $888.19
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-42-5 | |
104.15 | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
7501 | |
styrene |
C8H8 | |
MFCD00008612,MFCD00084450 | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
CHEBI:27452 | |
C=CC1=CC=CC=C1 |
Specifications
100-42-5,98-29-3 | |
0.9060g/mL | |
31°C | |
98.5% min. (GC) | |
C8H8 | |
C6H5CH=CH2 | |
10 mL | |
0.906 | |
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene | |
PPBRXRYQALVLMV-UHFFFAOYSA-N | |
styrene | |
10 to 20ppm p-tert-butylcatechol | |
0.695 mPa.s (25°C) | |
104.15 | |
99% | |
Liquid |
-31.0°C | |
145.0°C to 146.0°C | |
Authentic | |
Glass Bottle | |
1.5460 to 1.5480 | |
MFCD00008612,MFCD00084450 | |
05, 474 | |
15, 8990 | |
Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone,cs2,ccl4,dissolves organic substances and polymers | |
C=CC1=CC=CC=C1 | |
104.15 | |
7501 | |
CHEBI:27452 | |
7mbar at 20°C | |
Extra Pure | |
Styrene, Stabilized |
Safety and Handling
GHS P Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only