missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Styrene, 99%, extra pure, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 132790100

 View more versions of this product

Catalog No. AC132790100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Styrene
100-42-5,98-29-3
0.9060g/mL
31°C
98.5% min. (GC)
C8H8
C6H5CH=CH2
10 mL
0.906
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PPBRXRYQALVLMV-UHFFFAOYSA-N
styrene
10 to 20ppm p-tert-butylcatechol
0.695 mPa.s (25°C)
104.15
99%
Liquid
Stabilized
-31.0°C
145.0°C to 146.0°C
Authentic
Glass Bottle
1.5460 to 1.5480
MFCD00008612,MFCD00084450
05, 474
15, 8990
Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone,cs2,ccl4,dissolves organic substances and polymers
C=CC1=CC=CC=C1
104.15
7501
CHEBI:27452
7mbar at 20°C
Extra Pure

Chemical Identifiers

100-42-5
104.15
PPBRXRYQALVLMV-UHFFFAOYSA-N
7501
styrene
C8H8
MFCD00008612,MFCD00084450
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
CHEBI:27452
C=CC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure if inhaled.
May be fatal if swallowed and enters airways.<br/

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/prote

GHS Signal Word: Danger

EINECSNumber : 202-851-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only