missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinic Acid 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158740025

 View more versions of this product

Catalog No. AC158740025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Succinic acid
185.0°C to 190.0°C
235.0°C
C4H6O4
MFCD00002789
Solubility in water: 80g/L (20°C). Other solubilities: 1 kg/l boiling water, 1 g/18.5 mL alcohol - 1 g/6.3 mL methanol, 1 g/36 mL acetone - 1 g/20 mL glycerol, practically insoluble in petroleum ether, benzene,, carbon disulfide and carbon tetrachloride
OC(=O)CCC(O)=O
118.09
CHEBI:15741
99%
206°C
99%
110-15-6
99%
HO2CCH2CH2CO2H
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
KDYFGRWQOYBRFD-UHFFFAOYSA-N
butanedioic acid
1110
118.09
2.5 kg
Plastic bottle

Chemical Identifiers

110-15-6
118.09
KDYFGRWQOYBRFD-UHFFFAOYSA-N
1110
butanedioic acid
C4H6O4
MFCD00002789
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
CHEBI:15741
OC(=O)CCC(O)=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-740-4

RTECSNumber : WM4900000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only