Learn More
Succinic Acid 99%, Thermo Scientific Chemicals
$86.80 - $366.69
Chemical Identifiers
CAS | 110-15-6 |
---|---|
Molecular Formula | C4H6O4 |
Molecular Weight (g/mol) | 118.09 |
MDL Number | MFCD00002789 |
InChI Key | KDYFGRWQOYBRFD-UHFFFAOYSA-N |
Synonym | succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid |
PubChem CID | 1110 |
ChEBI | CHEBI:15741 |
IUPAC Name | butanedioic acid |
SMILES | OC(=O)CCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC158742500
|
Thermo Scientific Chemicals
158742500 |
250 g | Plastic bottle |
N/A
|
|
||||
AC158745000
|
Thermo Scientific Chemicals
158745000 |
500 g | Plastic bottle |
N/A
|
|
||||
AC158740010
|
Thermo Scientific Chemicals
158740010 |
1 kg | Plastic bottle |
N/A
|
|
||||
AC158740025
|
Thermo Scientific Chemicals
158740025 |
2.5 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-15-6 | |
118.09 | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
1110 | |
butanedioic acid |
C4H6O4 | |
MFCD00002789 | |
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
CHEBI:15741 | |
OC(=O)CCC(O)=O |
Specifications
185.0°C to 190.0°C | |
235.0°C | |
C4H6O4 | |
MFCD00002789 | |
Solubility in water: 80g/L (20°C). Other solubilities: 1 kg/l boiling water, 1 g/18.5 mL alcohol - 1 g/6.3 mL methanol, 1 g/36 mL acetone - 1 g/20 mL glycerol, practically insoluble in petroleum ether, benzene,, carbon disulfide and carbon tetrachloride | |
OC(=O)CCC(O)=O | |
118.09 | |
CHEBI:15741 | |
99% | |
206°C | |
Succinic acid, 99% |
110-15-6 | |
99% | |
HO2CCH2CH2CO2H | |
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
butanedioic acid | |
1110 | |
118.09 | |
250 g | |
Plastic bottle |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 203-740-4
RTECSNumber : WM4900000
TSCA : TSCA
RUO – Research Use Only