Learn More
Sulfolane, 99%, Thermo Scientific Chemicals
Miscible with water, acetone, toluene
$83.47 - $819.96
Chemical Identifiers
CAS | 126-33-0 |
---|---|
Molecular Formula | C4H8O2S |
Molecular Weight (g/mol) | 120.17 |
MDL Number | MFCD00005484 |
InChI Key | HXJUTPCZVOIRIF-UHFFFAOYSA-N |
Synonym | sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone |
PubChem CID | 31347 |
ChEBI | CHEBI:74794 |
SMILES | O=S1(=O)CCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1346630
|
Thermo Scientific Chemicals
A1346630 |
250 g |
Each for $83.47
|
|
||||||
AAA134660B
|
Thermo Scientific Chemicals
A134660B |
1000 g |
Each for $191.75
|
|
||||||
AAA134660I
|
Thermo Scientific Chemicals
A134660I |
5000 g |
Each for $819.96
|
|
||||||
Description
Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.
Solubility
Miscible with alcohol, water, acetone, toluene and dilute mineral acids. Slightly miscible with octanes, olefins and naphthenes.
Notes
Miscible with water, acetone, tolueneIncompatible with strong oxidizing agents.
Chemical Identifiers
126-33-0 | |
120.17 | |
HXJUTPCZVOIRIF-UHFFFAOYSA-N | |
31347 | |
O=S1(=O)CCCC1 |
C4H8O2S | |
MFCD00005484 | |
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone | |
CHEBI:74794 |
Specifications
126-33-0 | |
1.261 | |
166°C (330°F) | |
Weak, Characteristic | |
1.4825 | |
250 g | |
Hygroscopic | |
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone | |
HXJUTPCZVOIRIF-UHFFFAOYSA-N | |
1λâ¶-thiolane-1,1-dione | |
31347 | |
120.17 | |
Sulfolane |
25°C to 28°C | |
285°C | |
99% | |
C4H8O2S | |
MFCD00005484 | |
107765 | |
14,8955 | |
Miscible with alcohol,water,acetone,toluene and dilute mineral acids. Slightly miscible with octanes,olefins and naphthenes. | |
O=S1(=O)CCCC1 | |
120.17 | |
CHEBI:74794 | |
99% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 204-783-1
RTECSNumber : XN0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only