missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, 40% (v/v) Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S7284LTPL

 View more versions of this product

Catalog No. 18614268


Explore available promotions

Description

Description

Specifications

Specifications

7664-93-9,7732-18-5
Poly Bottle
4 L

Chemical Identifiers

7664-93-9
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.