missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 40% (v/v) Solution, Spectrum™ Chemical

$98.53 - $6158.62

Chemical Identifiers

CAS 7664-93-9
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614267
View Documents
Spectrum Chemical Mfg Cor
S7281LT
1 L Amber Glass Bottle
Each for $98.53
Only null left
 
18614268
View Documents
Spectrum Chemical Mfg Cor
S7284LTPL
4 L Amber Glass Bottle
Each for $280.74
Only null left
 
18614269
View Documents
Spectrum Chemical Mfg Cor
S728200LTBL
200 L Amber Glass Bottle
Each for $6,158.62
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7664-93-9

Specifications

7664-93-9,7732-18-5
Amber Glass Bottle
Ungraded
40%
1 L
Sulfuric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.