missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid (ACS Grade), Fisher Chemical™

Approx. Normality is 36

Supplier:  Thermo Fisher Scientific Formerly Fairlawn A300612GAL

 View more versions of this product

Catalog No. A300-612GAL


Explore available promotions

Description

Description

Specifications

Specifications

0.3
Colorless to Brown
7664-93-9
1.84g/cm³
Pass Test
3ppm max.
about 36
6.5 gal.
Liquid
1830
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
ACS Grade
Sulfuric Acid
10°C
290°C
0.8ppm max.
Glass Bottle
<0.001 mmHg at 20°C
95.0 to 98.0 w/w %
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
95.0 to 98.0% (w/w)

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes severe burns by all exposure routes. Reacts violently with water. Contact with combustible material may cause fire. Exposure to strong inorganic mists containing sulfuric acid may cause cancer by inhalation. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:2

DOTInformation : DOT Class 8, : Corrosive

SDS
Documents

Documents

Promotions

Promotions