Learn More
Sulfuric Acid (ACS Grade), Fisher Chemical™
Approx. Normality is 36
$181.38 - $6,223.62
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A300-500
|
Thermo Fisher Scientific Formerly Fairlawn
FLA300500 |
500 mL | Glass Bottle |
N/A
|
|
||||
A300S-500
|
Thermo Fisher Scientific Formerly Fairlawn
FLA300S500 |
500 mL | Safe-Cote™ Glass Bottle |
N/A
|
|
||||
A300-212
|
Thermo Fisher Scientific Formerly Fairlawn
FLA300212 |
2.5 L | Poly Bottle |
N/A
|
|
||||
A300SI-212
|
Thermo Fisher Scientific Formerly Fairlawn
A300SI212 |
2.5 L | Safe-Cote™ Glass Bottle |
N/A
|
|
||||
A300S-212
|
Thermo Fisher Scientific Formerly Fairlawn
FLA300S212 |
6 x 2.5 L | Safe-Cote™ Glass Bottle |
N/A
|
|
||||
A300-612GAL
|
Thermo Fisher Scientific Formerly Fairlawn
A300612GAL |
6.5 gal. | Glass Bottle |
N/A
|
|
||||
A300-225LB
|
Thermo Fisher Scientific Formerly Fairlawn
FLA300225LB |
225 lb. | Poly Drum |
N/A
|
|
||||
A300-700LB
|
Thermo Fisher Scientific Formerly Fairlawn
A300700LB |
700 lb. | Poly Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
0.3 | |
10°C | |
290°C | |
0.8ppm max. | |
Glass Bottle | |
<0.001 mmHg at °C | |
95.0 to 98.0 w/w % | |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
95.0 to 98.0% (w/w) | |
Sulfuric Acid |
Colorless to Brown | |
7664-93-9 | |
1.84g/cm³ | |
Pass Test | |
3ppm max. | |
about 36 | |
500 mL | |
Liquid | |
1830 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
ACS Grade |
Safety and Handling
DANGER!
Emergency Overview
Causes severe burns by all exposure routes. Reacts violently with water. Contact with combustible material may cause fire. Exposure to strong inorganic mists containing sulfuric acid may cause cancer by inhalation. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:3
Flammability:0
Instability:2
DOTInformation : DOT Class 8, : Corrosive