Learn More
Sulfuric Acid (ACS Grade), Fisher Chemical™
Approx. Normality is 36
Supplier: Thermo Fisher Scientific Formerly Fairlawn A300700LB
Specifications
0.3 | |
Colorless to Brown | |
7664-93-9 | |
1.84g/cm³ | |
Pass Test | |
3ppm max. | |
about 36 | |
700 lb. | |
Liquid | |
1830 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
ACS Grade |
Sulfuric Acid | |
10°C | |
290°C | |
0.8ppm max. | |
Poly Drum | |
<0.001 mmHg at 20°C | |
95.0 to 98.0 w/w % | |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
95.0 to 98.0% (w/w) |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
DANGER!
Emergency Overview
Causes severe burns by all exposure routes. Reacts violently with water. Contact with combustible material may cause fire. Exposure to strong inorganic mists containing sulfuric acid may cause cancer by inhalation. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:3
Flammability:0
Instability:2
DOTInformation : DOT Class 8, : Corrosive