missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid (Optima™), Fisher Chemical™

Supplier:  Seastar Chemicals Inc S020301TFIF03

 View more versions of this product

Catalog No. A468-500


Explore available promotions

Description

Description

  • Certificate of Lot Analysis included with each bottle.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture available on COA.
Specifications

Specifications

0.3
Colorless to Brown
7664-93-9
1.84g/cm³
<0.001 mmHg at 20°C
500 mL
Liquid
1830
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
OPTIMA™
Sulfuric Acid
10°C
290°C
FEP Bottle
Certificate of Lot Analysis included with each bottle.
H2O4S
MFCD00064589
15, 9104
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.07

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Accessories
Promotions

Promotions