missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfur powder, sublimed, -100 mesh, 99.5%, Thermo Scientific Chemicals
$116.70 - $281.23
Chemical Identifiers
CAS | 7704-34-9 |
---|---|
Molecular Formula | S |
Molecular Weight (g/mol) | 32.06 |
MDL Number | MFCD00085316 |
InChI Key | NINIDFKCEFEMDL-UHFFFAOYSA-N |
Synonym | polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot |
PubChem CID | 5362487 |
ChEBI | CHEBI:26833 |
IUPAC Name | sulfur |
SMILES | [S] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7704-34-9 | |
32.06 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
5362487 | |
sulfur |
S | |
MFCD00085316 | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
CHEBI:26833 | |
[S] |
Specifications
7704-34-9 | |
Sublimed Powder | |
99.50% | |
MFCD00085316 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
sulfur | |
5362487 | |
248°C (478°F) | |
Pale yellow powder |
Yellow | |
500 g | |
S | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
[S] | |
32.06 | |
CHEBI:26833 | |
Weak | |
Sulphur |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-722-6
RTECSNumber : WS4250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only