MilliporeSigma™ Supelco™ Supel™-Inert Multi-Layer Foil Gas Sampling Bag
Ideal for sampling low molecular weight compounds such as carbon monoxide (CO), carbon dioxide (CO2) and permanent gases
Supplier:  MilliporeSigma™ Supelco™ 30240U
 View more versions of this product
Catalog No. 111009920
Explore available promotions
Description
Description
Supel™-Inert multi-layer foil gas sampling bags with Thermogreen™ LB-2 septa are ideal for sampling low molecular weight compounds such as carbon monoxide (CO), carbon dioxide (CO2) and permanent gases. They are also suitable for sampling sulfur compounds such as hydrogen sulfide (H2S), carbon disulfide (CS2) and mercaptans. Key Features and Benefits: Multi-layer foil composed of two outer layers of aluminum film providing a great barrier to gases permeating through walls of the bag. Proven low-bleed Thermogreen™ LB-2 septa installed in the valve fitting. Chemically inert with moisture and light protection. Not recommended for low ppm VOCs due to background levels (we recommend the Supel™Inert PVDF Tedlar alternative film for VOCs). Two valve choices available: Screw Cap Valve (SCV) and Push/Pull Lock Valve (PLV). 4 sizes: 1 L, 2 L, 5 L, and 10 L
Specifications
Specifications
10 L
Multi-layer Foil
10/Pk.
Push/Pull Lock Valve (PLV)
Supel™
21 x 12 in.
Videos
SDS
Documents
Documents
Promotions
Promotions
Supel and Thermogreen are registered trademarks of Sigma-Aldrich Co. LLC