missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Supelco™ Supel™-Inert Multi-Layer Foil Gas Sampling Bag
Ideal for sampling low molecular weight compounds such as carbon monoxide (CO), carbon dioxide (CO2) and permanent gases
$220.18 - $363.17
Specifications
Includes | Push/Pull Lock Valve (PLV) |
---|---|
Material | Multi-layer Foil |
Product Line | Supel™ |
Quantity | 10/Pk. |
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W) | Price | Quantity | ||||
111010622
|
MilliporeSigma™ Supelco™
30281U |
0.6 L | 6 x 6 in. |
Each for $221.42
|
|
||||
111010596
|
MilliporeSigma™ Supelco™
30237U |
1 L | 9 x 7 in. |
Each for $220.18
|
|
||||
111010438
|
MilliporeSigma™ Supelco™
30238U |
2 L | 11 x 9 in. |
Each for $250.91
|
|
||||
111010106
|
MilliporeSigma™ Supelco™
30239U |
5 L | 14 x 12 in. |
Each for $286.37
|
|
||||
111009920
|
MilliporeSigma™ Supelco™
30240U |
10 L | 21 x 12 in. |
Each for $363.17
|
|
||||
Description
Supel™-Inert multi-layer foil gas sampling bags with Thermogreen™ LB-2 septa are ideal for sampling low molecular weight compounds such as carbon monoxide (CO), carbon dioxide (CO2) and permanent gases. They are also suitable for sampling sulfur compounds such as hydrogen sulfide (H2S), carbon disulfide (CS2) and mercaptans. Key Features and Benefits: Multi-layer foil composed of two outer layers of aluminum film providing a great barrier to gases permeating through walls of the bag. Proven low-bleed Thermogreen™ LB-2 septa installed in the valve fitting. Chemically inert with moisture and light protection. Not recommended for low ppm VOCs due to background levels (we recommend the Supel™Inert PVDF Tedlar alternative film for VOCs). Two valve choices available: Screw Cap Valve (SCV) and Push/Pull Lock Valve (PLV). 4 sizes: 1 L, 2 L, 5 L, and 10 LSpecifications
Push/Pull Lock Valve (PLV) | |
Multi-layer Foil | |
Supel™ | |
10/Pk. |
Supel and Thermogreen are registered trademarks of Sigma-Aldrich Co. LLC