Superior Glove™ Dexterity™ Nitrile Dipped Work Gloves
Seamless knit cotton shell is comfortable and flexible, with a special additive in the nitrile coating to make gloves 100% biodegradable.
Supplier: Superior Glove™ S15NT9
Description
- Nitrile dipped palm coating offers abrasion and puncture resistance
- An additive in the coating makes gloves 100% biodegradable
- Seamles knit cotton shell is comfortable and flexible
- CFIA compliant
Specifications
ANSI 4 | |
Nitrile | |
Specific | |
Dip-coated Gloves | |
Automotive Assembly, Maintenance, and Metal Stamping | |
ANSI/ISEA 105-16 Level 2 |
Seamless Knit | |
Blue, White | |
Cotton | |
9 | |
Knit | |
S15NT |