missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SuperSignal™ West Pico PLUS Chemiluminescent Substrate

Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.

$98.28 - $2985.42

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Product Line SuperSignal™
View More Specs

Products 6
Catalog Number Mfr. No. Kit Contents Quantity Price Quantity  
Catalog Number Mfr. No. Kit Contents Quantity Price Quantity  
PI34579
View Documents
Thermo Scientific™
34579
SuperSignal West Pico PLUS Luminol/Enhancer, SuperSignal West Pico PLUS Stable Peroxide 20 mL
Each for $98.28
 
PI34577
View Documents
Thermo Scientific™
34577
SuperSignal West Pico PLUS Luminol/Enhancer, SuperSignal West Pico PLUS Stable Peroxide 200 mL
Each for $322.17
 
PI34580
View Documents
Thermo Scientific™
34580
SuperSignal West Pico PLUS Luminol/Enhancer, SuperSignal West Pico PLUS Stable Peroxide 500 mL
Each for $559.17
 
FER34580X4
View Documents
Thermo Scientific™
34580X4
SuperSignal West Pico PLUS Luminol/Enhancer 4 x 500 mL
Each for $1,769.33
 
PI34578
View Documents
Thermo Scientific™
34578
SuperSignal West Pico PLUS Luminol/Enhancer, SuperSignal West Pico PLUS Stable Peroxide 1 L
Each for $908.33
 
FER34578X4
View Documents
Thermo Scientific™
34578X4
SuperSignal West Pico PLUS Luminol/Enhancer 4 x 1 L
Each for $2,985.42
 
Description

Description

Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.

SuperSignal West Pico PLUS Chemiluminescent Substrate characteristics include:

  • Sensitivity: pictogram to femtogram sensitivity
  • Stability: 8-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 6 to 24 hours
  • Recommended primary antibody concentration: 1:1000–1:5000 dilution (0.2–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

SuperSignal West Pico PLUS substrate is designed to provide excellent signal intensity and sensitivity for western blotting with HRP conjugates. The intensity of the light emission combined with the exceptional duration allows for the acquisition of multiple exposures to more easily obtain publication-quality blot images. SuperSignal West Pico PLUS substrate is compatible with different membranes, blocking reagents, and a wide range of antibody dilutions, making it an ideal choice for most western blotting applications.

Specifications

Specifications

Substrate
Liquid
SuperSignal™
Horseradish Peroxidase
6 to 24 hr.
Chemiluminescence
Western Blot
HRP
2°: 1:20-100 K (10-50 ng/mL), 1°: 1:1 K (0.2-1 μg/mL)
Room Temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.