Fisherbrand™ Superslip™ Cover Slips
Developed to minimize sticking, these cover slips are for use in automated coverslippers.
$11.91 - $19.71
Products 4
Catalog Number Mfr. No. Thickness (Metric) Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Thickness (Metric) Dimensions (L x W) Price Quantity  
12541054CA
View Documents
Fisherbrand™
24X401WYOO811AT
0.13 to 0.17 mm (#1) 40 x 24 mm
Pack for $16.51
Case of 10 Pack for $165.10
12541056CA
View Documents
Fisherbrand™
24X501WYOO811AT
0.13 to 0.17 mm (#1) 50 x 24 mm
Pack for $11.91
Case of 10 Pack for $119.10
12541055CA
View Documents
Fisherbrand™
24X5015WYO811AT
0.16 to 0.19 mm (#1.5) 50 x 24 mm
Pack for $19.71
Case of 10 Pack for $197.10
12541057CA
View Documents
Fisherbrand™
24X601WYOO811AT
0.13 to 0.17 mm (#1) 60 x 24 mm
Pack for $11.91
Case of 10 Pack for $119.10
Description
Description
All Superslip™ Cover Slips feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy.
  • Uniquely developed to reduce sticking
  • Can be used with automated coverslippers or for manual use in humid environments
  • Built-in moisture resistance
  • Manufactured with colorless borosilicate glass
  • Available in a variety of sizes and thicknesses
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'