missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
$235.06 - $324.50
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Format | Hinged Rack |
For Use With (Equipment) | Universal Fit |
Catalog Number | Mfr. No. | Volume (Metric) | Tip Style | Color | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Tip Style | Color | Price | Quantity | |||
02-707-439
![]() |
N/A | 0.1 to 10 μL | Micro Point | Natural |
Pack of 960 Each for $257.36
Case of 5 Pack for $1,286.80
|
![]() |
|||
02-707-442
|
N/A | 0.1 to 10 μL | Micro Point, Extended Length | Natural |
Pack of 960 Each for $266.54
Case of 5 Pack for $1,332.70
|
|
|||
02-707-470
|
N/A | 0.1 to 20 μL | Extended Length, Micro Point, Low Retention | Clear |
Pack of 960 Each for $292.82
Case of 5 Pack for $1,464.10
|
|
|||
02-707-432
|
N/A | 2 to 20 μL | Beveled | Natural |
Pack of 960 Each for $270.32
Case of 5 Pack for $1,351.60
|
|
|||
02-707-431
![]() |
Fisherbrand™
TF113100FIS |
10 to 100 μL | Beveled | Natural |
Pack of 960 Each for $235.06
Case of 5 Pack for $1,175.30
|
![]() |
|||
02-707-430
|
N/A | 20 to 200 μL | Micro Point | Natural |
Pack of 960 Each for $266.14
Case of 5 Pack for $1,330.70
|
|
|||
02-707-471
|
N/A | 30 to 300 μL | Micro Point | Clear |
Pack of 960 Each for $324.50
Case of 5 Pack for $1,622.50
|
|
|||
02-707-404
![]() |
N/A | 100 to 1000 μL | Micro Point | Natural |
Pack of 960 Each for $271.04
Case of 5 Pack for $1,355.20
|
![]() |
|||
Description
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Universal fit for research grade pipetters.
- Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
- DNase-/RNase-free; sterile tips are also pyrogen-free
- Sterile and clear color
- Filter eliminates aerosol cross-contamination without sealing or trapping samples
- Tips allow for noncontaminated sample recovery
- Not autoclavable
Specifications
Filtered | |
Not Autoclavable | |
Universal Fit |
Sterile | |
Hinged Rack | |
SureOne |