Thermo Scientific™ Spring Loaded Transfer Line Nut for ISQ and TSQ
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Supplier:  Thermo Scientific™ 1R1204340010
 View more versions of this product
Catalog No. 03-251-526
Explore available promotions
Description
Description
Specifications
Specifications
Thermo Scientific ISQ and TSQ
Spring Loaded Transfer Nut
Each