Thermo Scientific™ Spring Loaded Transfer Line Nut for ISQ and TSQ
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
$57.51 - $429.91
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
290BT240
|
Thermo Scientific™
290BT240 |
Chromatography Accessories | Swap-IT |
Pack of 5 Each for $57.51
|
|
||||
03-251-526
|
Thermo Scientific™
1R1204340010 |
Spring Loaded Transfer Nut | Thermo Scientific ISQ and TSQ |
Each for $429.91
|
|
||||