Learn More
sym-Diphenylcarbazide, 98%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158840250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
sym-Diphenylcarbazide | |
173.0°C to 176.0°C | |
1% max. | |
97.5 % min. (HPLC) | |
C13H14N4O | |
15,292 | |
15,3352 | |
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and acetone | |
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 | |
242.28 | |
CHEBI:4641 | |
98% | |
Powder or Flakes |
140-22-7 | |
Beige to Pink | |
Authentic | |
Glass bottle | |
25 g | |
(to chromate) passes test | |
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
KSPIHGBHKVISFI-UHFFFAOYSA-N | |
1,3-dianilinourea | |
8789 | |
242.28 | |
ACS Reagent |
Chemical Identifiers
140-22-7 | |
242.28 | |
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
CHEBI:4641 | |
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
C13H14N4O | |
KSPIHGBHKVISFI-UHFFFAOYSA-N | |
8789 | |
1,3-dianilinourea |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
GHS Signal Word: Warning
EINECSNumber : 205-403-7
RUO – Research Use Only