missing translation for 'onlineSavingsMsg'
Learn More

sym-Diphenylcarbazide, 98%, ACS reagent

CAS: 140-22-7 | C13H14N4O | 242.28 g/mol

$194.81 - $427.79

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.28
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158840250
SDS View Documents
Thermo Scientific Chemicals
158840250
25 g Glass bottle
Each for $194.81
Only null left
 
AC158841000
SDS View Documents
Thermo Scientific Chemicals
158841000
100 g Glass bottle
Each for $427.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

140-22-7
242.28
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
CHEBI:4641
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
C13H14N4O
KSPIHGBHKVISFI-UHFFFAOYSA-N
8789
1,3-dianilinourea

Specifications

140-22-7
Beige to Pink
Authentic
Glass bottle
25 g
(to chromate) passes test
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
KSPIHGBHKVISFI-UHFFFAOYSA-N
1,3-dianilinourea
8789
242.28
ACS Reagent
sym-Diphenylcarbazide
173.0°C to 176.0°C
1% max.
97.5 % min. (HPLC)
C13H14N4O
15,292
15,3352
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and acetone
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
242.28
CHEBI:4641
98%
Powder or Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g

GHS Signal Word: Warning

EINECSNumber : 205-403-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only