missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

sym-Diphenylcarbazide, 98%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158841000

 View more versions of this product

Catalog No. AC158841000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

sym-Diphenylcarbazide
173.0°C to 176.0°C
1% max.
97.5 % min. (HPLC)
C13H14N4O
15,292
15,3352
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and acetone
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
242.28
CHEBI:4641
98%
Powder or Flakes
140-22-7
Beige to Pink
Authentic
Glass bottle
100 g
(to chromate) passes test
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
KSPIHGBHKVISFI-UHFFFAOYSA-N
1,3-dianilinourea
8789
242.28
ACS Reagent

Chemical Identifiers

140-22-7
242.28
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
CHEBI:4641
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
C13H14N4O
KSPIHGBHKVISFI-UHFFFAOYSA-N
8789
1,3-dianilinourea
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g

GHS Signal Word: Warning

EINECSNumber : 205-403-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only