Synthware™ 4-Neck Round Bottom Flasks with Threaded Side Arm
For insertion of plan thermometers or bleed tubes having an O.D. between 5.0-7.0 mm
Supplier:  Synthware™ F714500
 View more versions of this product
Catalog No. 31501304
Explore available promotions
Description
Description
  • With standard taper outer joints and thermometer joints
  • Supplied complete with compression cap and Viton O-ring
Specifications
Specifications
Round bottom flask, 4-neck with threaded side arm
Borosilicate Glass (3.3)
Autoclavable
Rimmed
A
Glass Stopper
500 mL
Center: 24/40; 24/40 side
No
Non-sterile
Round
500 mL