Synthware™ Pressure Equalizing Funnel with Teflon™ Metering Stopcock
Teflon™ adjustable bore metering plug prevents sudden overflow, or non-flow during reagent addition.
Supplier: Synthware™ F611410
Description
- Fully control flow rate with the Teflon™ adjustable bore metering plug.
- Adjustment knob can be turned from full flow to dropwise.
- Prevents sudden overflow or non-flow during reagent addition.
Specifications
2 mm | |
Borosilicate Glass (3.3) | |
14/20 | |
Pressure-equalizing Funnel with Metering Stopcock |
10 mL | |
Clear | |
0.3 oz. |