Synthware™ Pressure Equalizing Funnel with Teflon™ Metering Stopcock
Teflon™ adjustable bore metering plug prevents sudden overflow, or non-flow during reagent addition.
$333.44 - $492.85
Specifications
Material Borosilicate Glass (3.3)
Color Clear
Product Type Pressure-equalizing Funnel with Metering Stopcock
View More Specs
Products 9
Catalog Number Mfr. No. Capacity (Metric) Joint Number Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Joint Number Price Quantity  
31501705
View Documents
Synthware™
F611410
10 mL 14/20
Each for $333.44
31501706
View Documents
Synthware™
F611425
25 mL 14/20
Each for $333.44
31502285
View Documents
Synthware™
F611925
25 mL 19/22
Each for $339.94
31501708
View Documents
Synthware™
F611960
60 mL 19/22
Each for $339.65
31501707
View Documents
Synthware™
F611460
60 mL 14/20
Each for $334.41
31501709
View Documents
Synthware™
F612460
60 mL 24/40
Each for $357.21
31501710
View Documents
Synthware™
F614125
125 mL 24/40
Each for $400.85
31501711
View Documents
Synthware™
F614250
250 mL 24/40
Each for $452.59
31501712
View Documents
Synthware™
F614500
500 mL 24/40
Each for $492.85
Description
Description
  • Fully control flow rate with the Teflon™ adjustable bore metering plug.
  • Adjustment knob can be turned from full flow to dropwise.
  • Prevents sudden overflow or non-flow during reagent addition.
Specifications
Specifications
Borosilicate Glass (3.3)
Clear
Pressure-equalizing Funnel with Metering Stopcock
SDS
missing translation for 'documents'
missing translation for 'documents'