Synthware™ Pressure Equalizing Funnel with Teflon™ Metering Stopcock
Teflon™ adjustable bore metering plug prevents sudden overflow, or non-flow during reagent addition.
Supplier:  Synthware™ F611960
 View more versions of this product
Catalog No. 31501708
Explore available promotions
Description
Description
  • Fully control flow rate with the Teflon™ adjustable bore metering plug.
  • Adjustment knob can be turned from full flow to dropwise.
  • Prevents sudden overflow or non-flow during reagent addition.
Specifications
Specifications
2 mm
Borosilicate Glass (3.3)
19/22
Pressure-equalizing Funnel with Metering Stopcock
60 mL
Clear
2 oz.