missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthware™ Coil Style Reflux Condenser

Coil length is 100mm in height

Supplier:  Synthware™ C351922

 View more versions of this product

Catalog No. 31500788


Explore available promotions

Description

Description

  • Condenser w/top outer joint set at 75° angle to body of condenser and lower inner drip tip
Specifications

Specifications

19/22
Reflux Condenser
Flask
Distillation
Glass
Videos
SDS
Documents

Documents

Promotions

Promotions