Synthware™ Coil Style Reflux Condenser
Coil length is 100mm in height
$276.41 - $286.50
Specifications
Material Glass
Type Reflux Condenser
For Use With (Application) Distillation
For Use With (Equipment) Flask
View More Specs
Products 2
Catalog Number Mfr. No. Standard Taper Joint No Price Quantity  
Catalog Number Mfr. No. Standard Taper Joint No Price Quantity  
31500787
View Documents
Synthware™
C351420
14/20
Each for $276.41
31500788
View Documents
Synthware™
C351922
19/22
Each for $286.50
Description
Description
  • Condenser w/top outer joint set at 75° angle to body of condenser and lower inner drip tip
Specifications
Specifications
Glass
Reflux Condenser
Distillation
Flask
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'