Synthware™ Distillation Adapter, 75°
Distillation adapter with standard taper inner joints on each end at a 75 deg. angle
Supplier: Synthware™ A152440
Description
Synthware distillation adapter bent at 75 degrees. Both ends are standard taper inner joints and are the same sizeSpecifications
Organic Synthesis | |
Glass Joint Adapter | |
24/40 |
Borosilicate Glass (3.3) | |
1 |