Synthware™ Reaction Flasks with High Vacuum Valve
With high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Vilton O-ring on shaft
Supplier: Synthware™ F549100
Description
SYNTHWARE round bottom single neck reaction flask with a standard taper outer joint and a high vacuum valve sidearm. Useful for sample storage or simple reactions. Valve plug may be removed to sample from the flask while an inert gas purge is passed through the sidearm.Specifications
Clear | |
Borosilicate Glass (3.3) | |
Reaction flask with high vacuum valve |
100 mL | |
Organic Synthesis | |
19/22 |