missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthware™ Reaction Flasks with High Vacuum Valve
With high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Vilton O-ring on shaft
$198.35 - $230.65
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass (3.3) |
For Use With (Application) | Organic Synthesis |
Product Type | Reaction flask with high vacuum valve |
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
31501413
|
Synthware™
F541050 |
50 mL | 14/20 |
Each for $198.35
|
|
||||
31501414
|
Synthware™
F541950 |
50 mL | 19/22 |
Each for $201.56
|
|
||||
31501415
|
Synthware™
F541100 |
100 mL | 14/20 |
Each for $198.35
|
|
||||
31501416
|
Synthware™
F549100 |
100 mL | 19/22 |
Each for $202.26
|
|
||||
31501417
|
Synthware™
F544100 |
100 mL | 24/40 |
Each for $201.91
|
|
||||
31501418
|
Synthware™
F541250 |
250 mL | 14/20 |
Each for $202.44
|
|
||||
31501419
|
Synthware™
F549250 |
250 mL | 19/22 |
Each for $208.76
|
|
||||
31501420
|
Synthware™
F544250 |
250 mL | 24/40 |
Each for $216.97
|
|
||||
31501421
|
Synthware™
F544500 |
500 mL | 24/40 |
Each for $221.85
|
|
||||
31501422
|
Synthware™
F54241L |
1000 mL | 24/40 |
Each for $230.65
|
|
||||
Description
SYNTHWARE round bottom single neck reaction flask with a standard taper outer joint and a high vacuum valve sidearm. Useful for sample storage or simple reactions. Valve plug may be removed to sample from the flask while an inert gas purge is passed through the sidearm.Specifications
Clear | |
Borosilicate Glass (3.3) | |
Organic Synthesis | |
Reaction flask with high vacuum valve |