Synthware™ Reducing Adapters
With standard taper joints at top and bottom
Supplier: Synthware™ A202429
Description
Synthware reducing adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is larger in size than the top outer joint.Specifications
Organic Synthesis | |
Reducing adapter | |
Top joint: 24/40; Bottom joint: 29/42 |
Borosilicate Glass (3.3) | |
1 |