Synthware™ Reflux Condenser
For use with volatile solvents
Supplier:  Synthware™ C264275
 View more versions of this product
Catalog No. 31500747
Explore available promotions
Description
Description
  • Coil style reflux condenser with tightly wrapped coil providing maximum cooling
  • With top outer standard taper joint and lower inner drip tip joint
Specifications
Specifications
Hose Connection: 19mm
None
Reflux Condenser
Borosilicate Glass (3.3)
24/40
Distillation
275 mm
Distillation Head