Synthware™ Round Bottom Flask with Threaded Inlet
Round bottom, single short neck flask with a standard taper outer joint
Supplier: Synthware™ F144100
Description
- Threaded side port for insertion of thermometer or bleed tube having O.D. between 5 mm and 7.5 mm
- Heavy wall design
- All sizes hand blown from tubing to ensure uniform wall thickness
Specifications
Joint: 24/40; With threaded inlet | |
100 mL |