Synthware™ Round Bottom Flask with Threaded Inlet
Round bottom, single short neck flask with a standard taper outer joint
$86.18 - $131.29
Products 8
Catalog Number Mfr. No. Description Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Description Capacity (Metric) Price Quantity  
31501143
View Documents
Synthware™
F141450
Joint: 14/20; With threaded inlet 50 mL
Each for $86.41
31501144
View Documents
Synthware™
F149100
Joint: 19/22; With threaded inlet 100 mL
Each for $103.94
31501142
View Documents
Synthware™
F141425
Joint: 14/20; With threaded inlet 25 mL
Each for $86.18
31501145
View Documents
Synthware™
F144100
Joint: 24/40; With threaded inlet 100 mL
Each for $115.76
31501146
View Documents
Synthware™
F141250
Joint: 14/20; With threaded inlet 250 mL
Each for $119.03
31501147
View Documents
Synthware™
F149250
Joint: 19/22; With threaded inlet 250 mL
Each for $121.15
31501148
View Documents
Synthware™
F144250
Joint: 24/40; With threaded inlet 250 mL
Each for $124.62
31501149
View Documents
Synthware™
F144500
Joint: 24/40; With threaded inlet 500 mL
Each for $131.29
Description
Description
  • Threaded side port for insertion of thermometer or bleed tube having O.D. between 5 mm and 7.5 mm
  • Heavy wall design
  • All sizes hand blown from tubing to ensure uniform wall thickness
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'