Synthware™ Round Bottom Flasks, 4-Neck
With standard taper outer joints on both the center and vertical side necks
Supplier:  Synthware™ F212500
 View more versions of this product
Catalog No. 31501310
Explore available promotions
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
Round bottom flask, 4-Neck
500 mL
Center: 29/42; 24/40 side